Skip to Content
Merck
Pseudoginsenoside F11 phyproof® Reference Substance

Pseudoginsenoside F11

Pseudoginsenoside F11 phyproof® Reference Substance
Synonym(s):
(3β,6α,12β,24R)-20,24-Epoxy-3,12,25-trihydroxydammaran-6-yl 2-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranoside, Ginsenoside A1
Empirical Formula (Hill Notation):
C42H72O14
CAS Number:
Molecular Weight:
801.01

Products

Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing