
Protoveratrine A

Empirical Formula (Hill Notation):
C41H63NO14
CAS Number:
Molecular Weight:
793.94
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing