![ProteoMass™ Cytochrome c MALDI-MS Standard vial of 10 nmol, (M+H+) 12,361.96 Da by calculation](/deepweb/assets/sigmaaldrich/product/images/143/853/fe0381c4-43d1-4192-aee0-e32bb344ff3d/640/fe0381c4-43d1-4192-aee0-e32bb344ff3d.jpg)
ProteoMass™ Cytochrome c MALDI-MS Standard
![ProteoMass™ Cytochrome c MALDI-MS Standard vial of 10 nmol, (M+H+) 12,361.96 Da by calculation](/deepweb/assets/sigmaaldrich/product/images/143/853/fe0381c4-43d1-4192-aee0-e32bb344ff3d/640/fe0381c4-43d1-4192-aee0-e32bb344ff3d.jpg)
Synonym(s):
Cytochrome c from equine heart, Cytochrome c from horse heart
CAS Number:
Products
Brand
Physical Form
Product Line
Quality Segment
Application
Package Size
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing