Skip to Content
Merck
Procyanidin A2 analytical standard

Procyanidin A2

Procyanidin A2 analytical standard
Synonym(s):
(+)-Epicatechin-(4β-8,2β-O-7)-epicatechin, Proanthocyanidin A2
Empirical Formula (Hill Notation):
C30H24O12
CAS Number:
Molecular Weight:
576.50

Products

Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing