
Procaine hydrochloride

Synonym(s):
4-Aminobenzoic acid 2-diethylaminoethyl ester, p-Aminobenzoic acid diethylaminoethyl ester hydrochloride, Novocaine hydrochloride
Linear Formula:
H2NC6H4CO2CH2CH2N(C2H5)2·HCl
CAS Number:
Molecular Weight:
272.77
Products
Product Category
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing