![Prasugrel for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/261/708/16a91396-2da7-41f9-a8cb-696a2d04999d/640/16a91396-2da7-41f9-a8cb-696a2d04999d.png)
Prasugrel for system suitability
![Prasugrel for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/261/708/16a91396-2da7-41f9-a8cb-696a2d04999d/640/16a91396-2da7-41f9-a8cb-696a2d04999d.png)
Synonym(s):
Prasugrel, 5-[2-Cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-4,5,6,7-tetrahydrothieno[3,2-c]pyridin-2-yl acetate, CS 747, CS-747, Effient, Efient, LY640315
Empirical Formula (Hill Notation):
C20H20FNO3S
CAS Number:
Molecular Weight:
373.44
Products
Formula Weight
Gene Alias
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing