Skip to Content
Merck
Potassium hexacyanoferrate(III) ACS reagent, ≥99.0%

Potassium hexacyanoferrate(III)

Potassium hexacyanoferrate(III) ACS reagent, ≥99.0%
Synonym(s):
Potassium ferricyanide, Red prussiate, Potassium hexacyanoferrate(III)
Linear Formula:
K3[Fe(CN)6]
CAS Number:
Molecular Weight:
329.24

Products

Product Category
Brand
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Reagent Type
Core
Special Grade