![Poly(ethylene glycol) dithiol average Mn 3,400](/deepweb/assets/sigmaaldrich/product/structures/396/978/61339a70-0465-4e2b-88a3-116e26d8a204/640/61339a70-0465-4e2b-88a3-116e26d8a204.png)
Poly(ethylene glycol) dithiol
![Poly(ethylene glycol) dithiol average Mn 3,400](/deepweb/assets/sigmaaldrich/product/structures/396/978/61339a70-0465-4e2b-88a3-116e26d8a204/640/61339a70-0465-4e2b-88a3-116e26d8a204.png)
Synonym(s):
PEG dithiol
Linear Formula:
HSCH2CH2(OCH2CH2)nSH
Products
Brand
Melting Point (°C)
Physical Form
Quality Segment
Functional Group
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Reagent Type
Showing 1-3 of 3