![Phenylbutyrate Related Compound C pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/187/434/bd50521b-a065-4aea-8bff-4320de52a0eb/640/bd50521b-a065-4aea-8bff-4320de52a0eb.png)
Phenylbutyrate Related Compound C
![Phenylbutyrate Related Compound C pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/187/434/bd50521b-a065-4aea-8bff-4320de52a0eb/640/bd50521b-a065-4aea-8bff-4320de52a0eb.png)
Synonym(s):
Cyclohexanebutyric acid, 4-Cyclohexylbutyric acid
Linear Formula:
C6H11(CH2)3CO2H
CAS Number:
Molecular Weight:
170.25
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2