![Pamabrom Related Compound A pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/202/124/00b6e323-1ebe-424e-b2ac-75738c81797b/640/00b6e323-1ebe-424e-b2ac-75738c81797b.png)
Pamabrom Related Compound A
![Pamabrom Related Compound A pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/202/124/00b6e323-1ebe-424e-b2ac-75738c81797b/640/00b6e323-1ebe-424e-b2ac-75738c81797b.png)
Synonym(s):
8-Chlorotheophylline, 8-Chloro-1,3-dimethyl-2,6(1H,3H)-purinedione
Empirical Formula (Hill Notation):
C7H7ClN4O2
CAS Number:
Molecular Weight:
214.61
Products
Brand
Formula Weight
Melting Point (°C)
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing