Skip to Content
Merck
O-[2-(3-Mercaptopropionylamino)ethyl]-O′-methylpolyethylene glycol 5,000

O-[2-(3-Mercaptopropionylamino)ethyl]-O′-methylpolyethylene glycol

O-[2-(3-Mercaptopropionylamino)ethyl]-O′-methylpolyethylene glycol 5,000
Synonym(s):
PEG-thiol, ‘Mercaptopolyethylene glycol monomethyl ether’
CAS Number:

Products

Brand
Quality Segment
Functional Group
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Reagent Type