Skip to Content
Merck
N-Methyl-N-propargyl-3-(2,4-dichlorophenoxy)propylamine hydrochloride ≥97% (GC)

N-Methyl-N-propargyl-3-(2,4-dichlorophenoxy)propylamine hydrochloride

N-Methyl-N-propargyl-3-(2,4-dichlorophenoxy)propylamine hydrochloride ≥97% (GC)
Synonym(s):
Clorgyline
Empirical Formula (Hill Notation):
C13H15Cl2NO · HCl
CAS Number:
Molecular Weight:
308.63
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Showing 1-1 of 1