![N-Malonyl-DL-tryptophan phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/191/684/3faecf53-6265-49fa-88d6-08f87a89b708/640/3faecf53-6265-49fa-88d6-08f87a89b708.png)
N-Malonyl-DL-tryptophan
![N-Malonyl-DL-tryptophan phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/191/684/3faecf53-6265-49fa-88d6-08f87a89b708/640/3faecf53-6265-49fa-88d6-08f87a89b708.png)
Synonym(s):
N-(2-Carboxyacetyl)-DL-tryptophan, Nα-Malonyl-DL-tryptophan
Empirical Formula (Hill Notation):
C14H14N2O5
Molecular Weight:
290.27
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing