Skip to Content
Merck
Neomycin trisulfate salt hydrate suitable for plant cell culture

Neomycin trisulfate salt hydrate

Neomycin trisulfate salt hydrate suitable for plant cell culture
Empirical Formula (Hill Notation):
C23H46N6O13 · 3H2SO4 · xH2O
CAS Number:
Molecular Weight:
908.88 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Physical Form
Product Line
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
Reference Material Type
Special Grade
Technique