![Monoclonal Anti-Cytokeratin Peptide 7 antibody produced in mouse clone LDS-68, ascites fluid](/deepweb/assets/sigmaaldrich/product/images/412/794/ed778d9a-1e62-4507-995c-781cea2332e1/640/ed778d9a-1e62-4507-995c-781cea2332e1.jpg)
Monoclonal Anti-Cytokeratin Peptide 7 antibody produced in mouse
![Monoclonal Anti-Cytokeratin Peptide 7 antibody produced in mouse clone LDS-68, ascites fluid](/deepweb/assets/sigmaaldrich/product/images/412/794/ed778d9a-1e62-4507-995c-781cea2332e1/640/ed778d9a-1e62-4507-995c-781cea2332e1.jpg)
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing