
Methyl valproate

Synonym(s):
2-Propylvaleric acid methyl ester, Methyl 2-propylpentanoate, Propylpentanoic acid methyl ester, Valproic acid methyl ester
Empirical Formula (Hill Notation):
C9H18O2
CAS Number:
Molecular Weight:
158.24
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing