Skip to Content
Merck
Methyl heptadecanoate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Methyl heptadecanoate

Methyl heptadecanoate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Heptadecanoic acid methyl ester, Methyl margarate
Linear Formula:
CH3(CH2)15COOCH3
CAS Number:
Molecular Weight:
284.48
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique