Skip to Content
Merck
Methyl cyclomyrcetone mixture of isomers, ≥90%

Methyl cyclomyrcetone

Methyl cyclomyrcetone mixture of isomers, ≥90%
Synonym(s):
7-Acetyl-1,2,3,4,5,6,7,8-octahydro-1,1,6,7-tetramethylnaphthalene, 1-(1,2,3,4,5,6,7,8-Octahydro-2,3,8,8-tetramethyl-2-naphthalenyl)ethanone, 2-Acetyl-2,3,8,8-tetramethyloctalin, Iso E Super
Empirical Formula (Hill Notation):
C16H26O
CAS Number:
Molecular Weight:
234.38

Products

Product Category
Brand
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing