Methotrexate Related Compound C
Synonym(s):
Methopterine, N10-Methylfolic acid
Empirical Formula (Hill Notation):
C20H21N7O6
CAS Number:
Molecular Weight:
455.42
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing