Metaxalone Related Compound C
Synonym(s):
Bis[2-hydroxy-3-(3,5-dimethylphenoxy)propyl]amine
Empirical Formula (Hill Notation):
C22H31NO4
CAS Number:
Molecular Weight:
373.49
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing