
Meloxicam Impurity C

Synonym(s):
N-[(2Z)-3,5-dimethylthiazol-2(3H)-ylidene]-4-hydroxy-2-methyl-2H-1,2- benzothiazine-3-carboxamide 1,1-dioxide
Empirical Formula (Hill Notation):
C15H15N3O4S2
Molecular Weight:
365.43
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing