![Medronic acid impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/331/546/b7ac1ed4-71f4-49c8-961e-fa4ca9a8711d/640/b7ac1ed4-71f4-49c8-961e-fa4ca9a8711d.png)
Medronic acid impurity A
![Medronic acid impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/331/546/b7ac1ed4-71f4-49c8-961e-fa4ca9a8711d/640/b7ac1ed4-71f4-49c8-961e-fa4ca9a8711d.png)
Synonym(s):
Triisopropyl phosphite
Linear Formula:
[(CH3)2CHO]3P
CAS Number:
Molecular Weight:
208.24
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing