![Meclocycline Sulfosalicylate pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/563/075/b0903072-6f04-46c1-8e4d-dd459c45ecf5/640/b0903072-6f04-46c1-8e4d-dd459c45ecf5.png)
Meclocycline sulfosalicylate
![Meclocycline Sulfosalicylate pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/563/075/b0903072-6f04-46c1-8e4d-dd459c45ecf5/640/b0903072-6f04-46c1-8e4d-dd459c45ecf5.png)
Synonym(s):
Meclocycline sulfosalicylate salt
Empirical Formula (Hill Notation):
C22H21ClN2O8 · C7H6O6S
CAS Number:
Molecular Weight:
695.05
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2