Skip to Content
Merck
Losartan ≥98% (HPLC)

Losartan

Losartan ≥98% (HPLC)
Synonym(s):
1-[1-[[2′-(2H-Tetrazol-5-yl)biphenyl-4-yl]methyl]-2-butyl-4-chloro-1H-imidazol-5-yl]methanol, 2-Butyl-4-chloro-1-[[2′-(1H-tetrazol-5-yl)-1,1′-biphenyl-4-yl]methyl]-1H-imidazole-5-methanol, 2-Butyl-4-chloro-1-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1H-imidazole-5-methanol, DUP 89
Empirical Formula (Hill Notation):
C22H23ClN6O
CAS Number:
Molecular Weight:
422.91

Products

Brand
Color
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1