![Loratadine Impurity H Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/350/238/45b036e0-0c5f-4a73-a293-0d7a5935f8db/640/45b036e0-0c5f-4a73-a293-0d7a5935f8db.png)
Loratadine impurity H
![Loratadine Impurity H Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/350/238/45b036e0-0c5f-4a73-a293-0d7a5935f8db/640/45b036e0-0c5f-4a73-a293-0d7a5935f8db.png)
Synonym(s):
Ethyl 4-oxo-1-piperidinecarboxylate, 1-Carbethoxy-4-piperidone, 1-Ethoxycarbonyl-4-piperidone
Empirical Formula (Hill Notation):
C8H13NO3
CAS Number:
Molecular Weight:
171.19
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2