![Lavandulol phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/222/812/53d3fee8-b96d-4a69-b2c2-274f7c1d97a7/640/53d3fee8-b96d-4a69-b2c2-274f7c1d97a7.png)
Lavandulol
![Lavandulol phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/222/812/53d3fee8-b96d-4a69-b2c2-274f7c1d97a7/640/53d3fee8-b96d-4a69-b2c2-274f7c1d97a7.png)
Synonym(s):
2-Isopropenyl-5-methyl-4-hexen-1-ol
Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing