![L-Aspartic acid monosodium salt monohydrate](/deepweb/assets/sigmaaldrich/product/structures/359/608/f314985c-a27d-4a22-9680-4f5a1512f5a2/640/f314985c-a27d-4a22-9680-4f5a1512f5a2.png)
L-Aspartic acid monosodium salt monohydrate
![L-Aspartic acid monosodium salt monohydrate](/deepweb/assets/sigmaaldrich/product/structures/359/608/f314985c-a27d-4a22-9680-4f5a1512f5a2/640/f314985c-a27d-4a22-9680-4f5a1512f5a2.png)
Synonym(s):
L-Aspartic acid sodium salt monohydrate, (S)-2-Aminobutanedioic acid sodium salt, Sodium L-aspartate
Linear Formula:
HO2CCH2CH(NH2)CO2Na · H2O
Products
Product Category
Brand
Formula Weight
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing