Skip to Content
Merck
Isovaleryl-L-carnitine ≥94.0% (HPLC)

Isovaleryl-L-carnitine

Isovaleryl-L-carnitine ≥94.0% (HPLC)
Synonym(s):
(2R)-3-Carboxy-N,N,N-trimethyl-2-(3-methyl-1-oxobutoxy)-1-propanaminium inner salt, L-Carnitine isovaleryl ester, iC5-Carnitine
Empirical Formula (Hill Notation):
C12H23NO4
CAS Number:
Molecular Weight:
245.32

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing