Iodine monobromide
Synonym(s):
Bromoiodine, Iodine monobromide
Linear Formula:
IBr
CAS Number:
Molecular Weight:
206.81
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing