![Imidurea Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/282/514/24d1e741-5587-401e-ac54-d1c8ec4f1c24/640/24d1e741-5587-401e-ac54-d1c8ec4f1c24.png)
Imidurea
![Imidurea Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/282/514/24d1e741-5587-401e-ac54-d1c8ec4f1c24/640/24d1e741-5587-401e-ac54-d1c8ec4f1c24.png)
Synonym(s):
Imidazolidinyl urea, N,N′′-Methylenebis[N′-[3-(hydroxymethyl)-2,5-dioxo-4-imidazolidinyl]-urea
Empirical Formula (Hill Notation):
C11H16N8O8
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing