Icariin
Synonym(s):
4′-O-methyl-8-γ,γ-dimethylallylkaempferol-3-rhamnoside-7-glucoside
Empirical Formula (Hill Notation):
C33H40O15
CAS Number:
Molecular Weight:
676.66
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing