Skip to Content
Merck
Hydroxylamine sulfate 99.999% trace metals basis

Hydroxylamine sulfate

Hydroxylamine sulfate 99.999% trace metals basis
Synonym(s):
Hydroxylammonium sulfate
Linear Formula:
(NH2OH)2 · H2SO4
CAS Number:
Molecular Weight:
164.14
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing