Skip to Content
Merck
Hexyl alcohol FCC, FG

Hexyl alcohol

Hexyl alcohol FCC, FG
Synonym(s):
1-Hexanol, Hexyl alcohol
Linear Formula:
CH3(CH2)5OH
CAS Number:
Molecular Weight:
102.17
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing