Skip to Content
Merck
Heneicosane analytical standard

Heneicosane

Heneicosane analytical standard
Linear Formula:
CH3(CH2)19CH3
CAS Number:
Molecular Weight:
296.57

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing