Skip to Content
Merck
Hamamelitannin phyproof® Reference Substance

Hamamelitannin

Hamamelitannin phyproof® Reference Substance
Synonym(s):
2-C-(Hydroxymethyl)-D-ribofuranose 2′,5-digallate, Hamamelofuranose 2′,5-digallate
Empirical Formula (Hill Notation):
C20H20O14
CAS Number:
Molecular Weight:
484.36
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing