Skip to Content
Merck
Guanosine 5′-triphosphate sodium salt hydrate ≥95% (HPLC), powder

Guanosine 5′-triphosphate sodium salt hydrate

Guanosine 5′-triphosphate sodium salt hydrate ≥95% (HPLC), powder
Synonym(s):
GTP
Empirical Formula (Hill Notation):
C10H16N5O14P3 · xNa+
CAS Number:
Molecular Weight:
523.18 (free acid basis)
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing