Skip to Content
Merck
Glycerol-d8 ≥98 atom % D, ≥98% (CP)

Glycerol-d8

Glycerol-d8 ≥98 atom % D, ≥98% (CP)
Synonym(s):
1,2,3-Propanetriol-d8, Deuterated glycerol, Glycerin-d8
Linear Formula:
(DOCD2)2CDOD
CAS Number:
Molecular Weight:
100.14

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing