![Glutathione Impurity A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/414/402/c72a531d-113d-436a-9332-1fac08ef9108/640/c72a531d-113d-436a-9332-1fac08ef9108.png)
Glutathione Impurity A
![Glutathione Impurity A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/414/402/c72a531d-113d-436a-9332-1fac08ef9108/640/c72a531d-113d-436a-9332-1fac08ef9108.png)
Synonym(s):
Cys-Gly
Empirical Formula (Hill Notation):
C5H10N2O3S
CAS Number:
Molecular Weight:
178.21
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing