
GBR 12909 dihydrochloride

Synonym(s):
1-(2-[bis(4-Fluorophenyl)methoxy]ethyl)-4-(3-phenylpropyl)piperazine dihydrochloride, GBR 12909 Dihydrochloride
Empirical Formula (Hill Notation):
C28H32F2N2O · 2HCl
CAS Number:
Molecular Weight:
523.49
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing