![Furanyl fentanyl hydrochloride solution 100 μg/mL in methanol (as free base), ampule of 0.5 mL, certified reference material, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/388/343/4cd533e3-c6e8-4a38-a74e-adb659052807/640/4cd533e3-c6e8-4a38-a74e-adb659052807.png)
Furanyl fentanyl hydrochloride solution
![Furanyl fentanyl hydrochloride solution 100 μg/mL in methanol (as free base), ampule of 0.5 mL, certified reference material, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/388/343/4cd533e3-c6e8-4a38-a74e-adb659052807/640/4cd533e3-c6e8-4a38-a74e-adb659052807.png)
Empirical Formula (Hill Notation):
C24H26N2O2 · HCl
Molecular Weight:
410.94
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing