![Fluoxetine impurity A CRS, European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/419/170/f2508b5a-4b91-472d-a5ae-103d795279bc/640/f2508b5a-4b91-472d-a5ae-103d795279bc.png)
Fluoxetine impurity A
![Fluoxetine impurity A CRS, European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/419/170/f2508b5a-4b91-472d-a5ae-103d795279bc/640/f2508b5a-4b91-472d-a5ae-103d795279bc.png)
Synonym(s):
α-[2-(Methylamino)ethyl]benzyl alcohol, 3-(Methylamino)-1-phenylpropan-1-ol
Linear Formula:
C6H5CH(CH2CH2NHCH3)OH
CAS Number:
Molecular Weight:
165.23
Products
Formula Weight
Manufacturer
Melting Point (°C)
Markush Class
Markush Group
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing