Skip to Content
Merck
Fluorescein-12-UTP ≥85% (HPLC), solution

Fluorescein-12-UTP

Fluorescein-12-UTP ≥85% (HPLC), solution
Synonym(s):
Fluorescein-12-UTP tetralithium salt, Fluorescein-5(6)-carboxamidocaproyl-(5-[3-aminoallyl]uridine 5′-triphosphate)
Empirical Formula (Hill Notation):
C39H41N4O22P3
CAS Number:
Molecular Weight:
1010.68

Products

Formula Weight
Manufacturer
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing