![Fast Red TR Salt Dye content 15 %](/deepweb/assets/sigmaaldrich/product/structures/129/786/c74e6df1-dea6-411e-93bd-7d4e3e720e0b/640/c74e6df1-dea6-411e-93bd-7d4e3e720e0b.png)
Fast Red TR Salt
![Fast Red TR Salt Dye content 15 %](/deepweb/assets/sigmaaldrich/product/structures/129/786/c74e6df1-dea6-411e-93bd-7d4e3e720e0b/640/c74e6df1-dea6-411e-93bd-7d4e3e720e0b.png)
Synonym(s):
Fast Red TR Salt hemi(zinc chloride) salt, 4-Chloro-2-methylbenzenediazonium salt, Azoic Diazo No. 11
Empirical Formula (Hill Notation):
C7H6Cl2N2 · 0.5 ZnCl2
CAS Number:
Molecular Weight:
257.19
Products
Product Category
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing