Etravirine System Suitability Mixture
Synonym(s):
4-((6-Amino-5-bromo-2-((4-cyanophenyl)amino)pyrimidin-4-yl)oxy)-3,5-dimethylbenzonitrile
Empirical Formula (Hill Notation):
C20H15BrN6O
CAS Number:
Molecular Weight:
435.28
Products
Formula Weight
Manufacturer
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing