Skip to Content
Merck
Etoricoxib ≥95% (HPLC)

Etoricoxib

Etoricoxib ≥95% (HPLC)
Synonym(s):
5-Chloro-3-[4-(methylsulfonyl)phenyl]-2-(2-methyl-5-pyridinyl)pyridine, 5-Chloro-6′-methyl-3-[4-(methylsulfonyl)phenyl]-2,3′-bipyridine
Empirical Formula (Hill Notation):
C18H15ClN2O2S
CAS Number:
Molecular Weight:
358.84

Products

Brand
Color
Formula Weight
Gene Alias
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing