Skip to Content
Merck
Dimethyl isosorbide BioRenewable, anhydrous, DMF and NMP substitute, ≥99%

Dimethyl isosorbide

Dimethyl isosorbide BioRenewable, anhydrous, DMF and NMP substitute, ≥99%
Synonym(s):
Isosorbide dimethyl ether, 1,4:3,6-Dianhydro-2,5-di-O-methyl-D-glucitol, 1,4:3,6-Dianhydrosorbitol 2,5-dimethyl ether, 2,5-Di-O-methyl-1,4:3,6-dianhydro-D-glucitol, DMI
Empirical Formula (Hill Notation):
C8H14O4
CAS Number:
Molecular Weight:
174.19

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Color
Feature
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group