
Dienogest for system suitability

Synonym(s):
Dienogest, (17a)-17-Hydroxy-3-oxo-19-norpregna-4,9-diene-21-nitrile, 17-Hydroxy-3-oxo-19-norpregna-4,9-diene-21-nitrile, 17-cyanomethyl-17-hydroxy-estra-4,9-dien-3-one
Empirical Formula (Hill Notation):
C20H25NO2
CAS Number:
Molecular Weight:
311.42
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing