Skip to Content
Merck
Dibutyltin dilaurate 95%

Dibutyltin dilaurate

Dibutyltin dilaurate 95%
Synonym(s):
Dibutyltin dilaurate
Linear Formula:
(CH3CH2CH2CH2)2Sn[OCO(CH2)10CH3]2
CAS Number:
Molecular Weight:
631.56

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing