![D-Camphor Impurity F, Fenchol Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/232/077/ac327e75-9811-4d0b-8c7e-8cf486e137c4/640/ac327e75-9811-4d0b-8c7e-8cf486e137c4.png)
D-Camphor Impurity F, Fenchol
![D-Camphor Impurity F, Fenchol Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/232/077/ac327e75-9811-4d0b-8c7e-8cf486e137c4/640/ac327e75-9811-4d0b-8c7e-8cf486e137c4.png)
Synonym(s):
(+)-Fenchol, (1R)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-ol, Fenchyl alcohol
Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing