Skip to Content
Merck
Cortisone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Cortisone

Cortisone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
17α,21-Dihydroxy-4-pregnene-3,11,20-trione, 17α-Hydroxy-11-dehydrocorticosterone, 4-Pregnene-17α,21-diol-3,11,20-trione, Kendall’s Compound E, Reichstein’s Substance Fa, Cortisone
Empirical Formula (Hill Notation):
C21H28O5
CAS Number:
Molecular Weight:
360.44
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Sterility
Technique